Assay Details
| Gene Symbol: Ssb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031119.2 | NP_112381.2 | 7 - 8 | 728 | 76 | |
| XM_006234321.2 | XP_006234383.1 | 7 - 8 | 804 | 76 | ||
| XM_006234322.2 | XP_006234384.1 | 7 - 8 | 804 | 76 | ||
| XM_006234323.2 | XP_006234385.1 | 7 - 8 | 804 | 76 | ||
| GenBank mRNA | BC081780.1 | - | 7 - 8 | 728 | 7192454 | 76 |
| BC097252.1 | - | 6 - 7 | 808 | 76 | ||
| X67859.1 | - | 6 - 7 | 663 | 76 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ssb
Genomic Map