Assay Details
| Gene Symbol: Foxm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031633.3 | NP_113821.2 | 3 - 4 | 733 | 86 | |
| XM_006237416.2 | XP_006237478.1 | 2 - 3 | 709 | 86 | ||
| XM_006237417.2 | XP_006237479.1 | 2 - 3 | 709 | 86 | ||
| XM_006237418.2 | XP_006237480.1 | 3 - 4 | 733 | 86 | ||
| XM_006237419.2 | XP_006237481.1 | 2 - 3 | 709 | 86 | ||
| XM_006237420.2 | XP_006237482.1 | 3 - 4 | 733 | 86 | ||
| XM_006237421.2 | XP_006237483.1 | 3 - 4 | 733 | 86 | ||
| GenBank mRNA | U83112.1 | - | 3 - 4 | 733 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Foxm1
Genomic Map