Assay Details
| Gene Symbol: Klrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031649.2 | NP_113837.1 | 3 - 4 | 357 | 69 | |
| XM_006237313.2 | XP_006237375.1 | 2 - 3 | 320 | 69 | ||
| GenBank mRNA | X79812.1 | - | 3 - 4 | 410 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Klrg1
Genomic Map