Assay Details
| Gene Symbol: Slco1b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270586.1 | NP_001257515.1 | 13 - 14 | 1743 | 105 | |
| NM_001270587.1 | NP_001257516.1 | 12 - 13 | 1749 | 105 | ||
| NM_031650.3 | NP_113838.1 | 13 - 14 | 1848 | 105 | ||
| GenBank mRNA | AF147740.1 | - | 12 - 13 | 1629 | 105 | |
| AF208545.2 | - | 13 - 14 | 1848 | 105 | ||
| AF217450.2 | - | 12 - 13 | 1749 | 105 | ||
| AJ271682.1 | - | 13 - 14 | 1849 | 105 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slco1b2
Genomic Map