Assay Details
| Gene Symbol: Gcs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031749.2 | NP_113937.1 | 1 - 2 | 346 | 91 | |
| GenBank mRNA | AF087431.1 | - | 1 - 2 | 347 | 91 | |
| Gene Symbol: LOC103692171 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008763086.1 | XP_008761308.1 | 414 | 91 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gcs1
Genomic Map