Assay Details
Gene Symbol: Flot2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270800.1 | NP_001257729.1 | 4 - 5 | 448 | 100 | |
NM_001270801.1 | NP_001257730.1 | 4 - 5 | 457 | 100 | ||
NM_031830.2 | NP_114018.1 | 4 - 5 | 448 | 100 | ||
NR_073076.1 | 3 - 4 | 366 | 100 | |||
NR_073077.1 | 3 - 4 | 357 | 100 | |||
XM_006246952.1 | XP_006247014.1 | 5 - 6 | 1069 | 100 | ||
XM_008768119.1 | XP_008766341.1 | 3 - 4 | 435 | 100 | ||
GenBank mRNA | AF023302.1 | - | 4 - 5 | 427 | 100 | |
AF023303.1 | - | 4 - 5 | 427 | 100 | ||
AF023304.1 | - | 3 - 4 | 345 | 100 | ||
BC083550.1 | - | 3 - 4 | 324 | 7133999 | 100 | |
CB581049.1 | - | 4 - 5 | 448 | 100 | ||
U64999.1 | - | 1 - 2 | 300 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
