Assay Details
Gene Symbol: LOC100910021 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750567.3 | XP_003750615.1 | 4 - 5 | 951 | 109 |
Gene Symbol: Pik3cb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006243642.2 | XP_006243704.1 | 21 - 22 | 3092 | 109 | |
XM_008766567.1 | XP_008764789.1 | 16 - 17 | 2479 | 109 | ||
GenBank mRNA | AJ012482.1 | - | 21 - 22 | 3075 | 109 | |
CV117575.1 | - | 1 - 2 | 183 | 109 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cell-matrix adhesion
cellular calcium ion homeostasis
endocytosis
autophagy
cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
platelet activation
phosphatidylinositol-3-phosphate biosynthetic process
embryonic cleavage
phosphatidylinositol phosphorylation
phosphatidylinositol-mediated signaling
ATP binding
1-phosphatidylinositol-3-kinase activity
phosphatidylinositol 3-kinase activity
1-phosphatidylinositol-4-phosphate 3-kinase activity
insulin receptor substrate binding
phosphatidylinositol-4,5-bisphosphate 3-kinase activity
regulation of cell-matrix adhesion
cellular calcium ion homeostasis
endocytosis
autophagy
cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
platelet activation
phosphatidylinositol-3-phosphate biosynthetic process
embryonic cleavage
phosphatidylinositol phosphorylation
phosphatidylinositol-mediated signaling
ATP binding
1-phosphatidylinositol-3-kinase activity
phosphatidylinositol 3-kinase activity
1-phosphatidylinositol-4-phosphate 3-kinase activity
insulin receptor substrate binding
phosphatidylinositol-4,5-bisphosphate 3-kinase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P088841 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P12820 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 689636 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 455205 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 761464 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 777143 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899979 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683639 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 502242 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27319 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 407197 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 142350 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586980 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 150808 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 623658 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 532038 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 265965 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687901 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 133308 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 822815 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 425573 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 541153 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138602 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 762916 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 486285 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 197061 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 578718 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 19850 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 454335 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 957671 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 139664 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 585028 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 364664 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571695 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 728861 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 540857 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 842084 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 357465 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 561744 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 643373 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763895 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 843073 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69690 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448074 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27893 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731695 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 466972 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 179362 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741317 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383967 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920868 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 495612 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 630433 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 445761 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 679989 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 757885 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 843339 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 620691 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 57145 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 388685 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 134059 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 108946 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 113951 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 31378 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 81077 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 90672 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 43399 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 90037 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 21397 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 67228 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P12820 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P377432 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P088841 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P088836 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P158035 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P088841 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P12820 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P158035 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P088836 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P377432 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P377432 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P088836 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P158035 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P088841 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P12820 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P104679 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P12820 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 101352 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 24713 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 102589 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 94503 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 71874 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 23145 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 41426 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10919466 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10919461 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10919469 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10820199 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10820194 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10919460 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10820166 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10820168 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10820165 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10919456 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10820200 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10820164 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10919457 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10919454 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10820191 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10820202 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10820197 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10820196 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10919462 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10820201 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10820193 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10919463 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10919465 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10820203 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10820195 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10919455 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10919459 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10820167 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10919467 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10919458 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10820198 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10820204 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10820205 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10919468 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10820170 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10820163 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10919453 | - | - |
Rat Genome 230 2.0 Array | 1377145_PM_at | - | - |
Rat Genome 230 2.0 Array | 1377145_at | - | - |
Rat Genome 230 2.0 Array | 1387847_PM_at | - | - |
Rat Genome 230 2.0 Array | 1387847_at | - | - |
Affymetrix HT Rat Focus Array | 1383799_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1387847_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1387847_at | - | - |
Affymetrix HT Rat Focus Array | 1383799_at | - | - |
Rat Expression Set 230 | 1387847_at | - | - |
Rat Expression Set 230 | 1387847_PM_at | - | - |
Rat Expression Set 230 | 1377145_at | - | - |
Rat Expression Set 230 | 1377145_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10820170 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10919453 | - | - |
Rat Genome U34 Set | AJ012482_at | - | - |
Rat Genome U34 Set | rc_AA819646_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20718184 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1368644 | - | - |
RatRef-12 Expression BeadChip | ILMN_66637 | - | - |
