Assay Details
| Gene Symbol: Sc5d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053642.2 | NP_446094.1 | 3 - 4 | 341 | 107 | |
| GenBank mRNA | AB052846.1 | - | 2 - 3 | 319 | 107 | |
| BC081704.1 | - | 3 - 4 | 341 | 7131154 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sc5d
Genomic Map