Assay Details
| Gene Symbol: Kidins220 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053795.1 | NP_446247.1 | 8 - 9 | 908 | 61 | |
| XM_006239923.2 | XP_006239985.1 | 7 - 8 | 1307 | 61 | ||
| XM_006239924.2 | XP_006239986.1 | 7 - 8 | 1304 | 61 | ||
| XM_006239925.2 | XP_006239987.1 | 8 - 9 | 997 | 61 | ||
| XM_006239926.2 | XP_006239988.1 | 7 - 8 | 1307 | 61 | ||
| XM_006239927.2 | XP_006239989.1 | 8 - 9 | 994 | 61 | ||
| XM_006239928.2 | XP_006239990.1 | 8 - 9 | 994 | 61 | ||
| XM_006239929.2 | XP_006239991.1 | 7 - 8 | 1307 | 61 | ||
| XM_006239931.2 | XP_006239993.1 | 7 - 8 | 1307 | 61 | ||
| XM_006239932.2 | XP_006239994.1 | 7 - 8 | 1307 | 61 | ||
| XM_006239933.2 | XP_006239995.1 | 7 - 8 | 1306 | 61 | ||
| XM_006239934.2 | XP_006239996.1 | 7 - 8 | 1306 | 61 | ||
| XM_006239935.2 | XP_006239997.1 | 7 - 8 | 1306 | 61 | ||
| XM_006239936.2 | XP_006239998.1 | 7 - 8 | 1306 | 61 | ||
| XM_006239937.2 | XP_006239999.1 | 8 - 9 | 997 | 61 | ||
| GenBank mRNA | AF239045.2 | - | 8 - 9 | 908 | 61 | |
| AF313464.1 | - | 7 - 8 | 800 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kidins220
Genomic Map