Assay Details
| Gene Symbol: Mprip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034022.1 | NP_001029194.1 | 16 - 17 | 2404 | 77 | |
| NM_053814.2 | NP_446266.2 | 17 - 18 | 2512 | 77 | ||
| XM_006246422.2 | XP_006246484.1 | 18 - 19 | 6938 | 77 | ||
| XM_006246423.2 | XP_006246485.1 | 18 - 19 | 6938 | 77 | ||
| XM_006246424.2 | XP_006246486.1 | 18 - 19 | 6938 | 77 | ||
| XM_006246425.2 | XP_006246487.1 | 18 - 19 | 6583 | 77 | ||
| XM_006246427.2 | XP_006246489.1 | 17 - 18 | 6476 | 77 | ||
| XM_006246430.2 | XP_006246492.1 | 12 - 13 | 8713 | 77 | ||
| XM_006246431.2 | XP_006246493.1 | 17 - 18 | 3121 | 77 | ||
| XM_006246432.2 | XP_006246494.1 | 17 - 18 | 2765 | 77 | ||
| XM_008767759.1 | XP_008765981.1 | 17 - 18 | 6869 | 77 | ||
| XM_008767760.1 | XP_008765982.1 | 17 - 18 | 6830 | 77 | ||
| XM_008767761.1 | XP_008765983.1 | 16 - 17 | 6762 | 77 | ||
| XM_008767762.1 | XP_008765984.1 | 16 - 17 | 3004 | 77 | ||
| GenBank mRNA | AF311311.1 | - | 17 - 18 | 2442 | 77 | |
| BC098911.1 | - | 16 - 17 | 2390 | 7446349 | 77 | |
| FQ222001.1 | - | 17 - 18 | 2904 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mprip
Genomic Map