Assay Details
Gene Symbol: Gucy2f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053831.1 | NP_446283.1 | 17 - 18 | 3404 | 120 | |
XM_008773386.1 | XP_008771608.1 | 16 - 17 | 3147 | 120 | ||
GenBank mRNA | L36030.1 | - | 17 - 18 | 3404 | 120 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P088750 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 722641 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 761646 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 599551 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 103482 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 97089 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144443 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 300326 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379942 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 499203 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115437 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 81813 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 612264 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304045 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202851 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106750 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 949710 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 157494 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 726382 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496851 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 924659 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 839762 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621184 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95294 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 126860 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 342060 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 702726 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 645149 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205536 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 906064 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334699 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 649601 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 724009 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 238882 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617659 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 274018 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 319536 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224868 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 671626 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939952 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 179988 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799173 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 426577 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 149010 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 555767 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 844094 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 937560 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 107560 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 605819 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 548130 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562422 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 702475 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 671614 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 340715 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 514444 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 241264 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 420759 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744139 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 866466 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681597 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422667 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962491 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 100462 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 973530 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 847652 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 884555 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 26730 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 451427 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153618 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 456672 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 151983 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 145957 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 212839 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763400 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 130940 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 211230 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 327120 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 491838 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 14083 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 31180 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 74382 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 123055 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 28195 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 35527 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 94358 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 39175 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 117740 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145601 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P088750 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P088750 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P088750 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P179824 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 35766 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 41969 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 104907 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 80535 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 90495 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 48084 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 10122 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 50282 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 31660 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10932823 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10932826 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10932829 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10932835 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10932820 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10932816 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10932833 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10932821 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10932827 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10932824 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10932832 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10932822 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10932828 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10932818 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10932830 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10932825 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10932819 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10932817 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10932834 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10932831 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10932815 | - | - |
Rat Genome 230 2.0 Array | 1369356_at | - | - |
Rat Genome 230 2.0 Array | 1369356_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1369356_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1369356_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0X00014225.rn.1 | TC0X00002270.rn.1 | RTA TC |
Rat Expression Set 230 | 1369356_at | - | - |
Rat Expression Set 230 | 1369356_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10932815 | - | - |
Rat Genome U34 Set | L36030_at | - | - |
Rat Genome U34 Set | L36030_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21936942 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1363122 | - | - |
RatRef-12 Expression BeadChip | ILMN_53180 | - | - |
