Assay Details
| Gene Symbol: Eltd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022294.1 | NP_071630.1 | 9 - 10 | 1456 | 89 | |
| GenBank mRNA | AF192401.1 | - | 9 - 10 | 1456 | 89 | |
| AF192402.1 | - | 9 - 10 | 1456 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Eltd1
Genomic Map