Assay Details
| Gene Symbol: Qdpr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022390.1 | NP_071785.1 | 3 - 4 | 336 | 80 | |
| GenBank mRNA | BC072536.1 | - | 3 - 4 | 349 | 7103182 | 80 |
| FQ213296.1 | - | 3 - 4 | 315 | 80 | ||
| FQ213843.1 | - | 3 - 4 | 366 | 80 | ||
| J03481.1 | - | 3 - 4 | 336 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Qdpr
Genomic Map