Assay Details
Gene Symbol: Grk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022928.1 | NP_075217.1 | 12 - 13 | 1456 | 67 | |
XM_006251359.1 | XP_006251421.1 | 11 - 12 | 1199 | 67 | ||
XM_006251360.1 | XP_006251422.1 | 12 - 13 | 1292 | 67 | ||
XM_008770353.1 | XP_008768575.1 | 12 - 13 | 1292 | 67 | ||
XM_008770354.1 | XP_008768576.1 | 11 - 12 | 1199 | 67 | ||
XM_008770355.1 | XP_008768577.1 | 12 - 13 | 1292 | 67 | ||
XM_008770356.1 | XP_008768578.1 | 8 - 9 | 803 | 67 | ||
XM_008770357.1 | XP_008768579.1 | 7 - 8 | 847 | 67 | ||
XM_008770358.1 | XP_008768580.1 | 5 - 6 | 1401 | 67 | ||
GenBank mRNA | X97568.1 | - | 12 - 13 | 1456 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
