Assay Details
| Gene Symbol: Gucy1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023956.1 | NP_076446.1 | 4 - 5 | 1620 | 81 | |
| GenBank mRNA | AB079780.1 | - | 1 - 2 | 46 | 81 | |
| AB096080.1 | - | 4 - 5 | 1620 | 81 | ||
| AB097860.1 | - | 4 - 5 | 1200 | 81 | ||
| AF109963.2 | - | 4 - 5 | 1620 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P154855 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P369825 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 706977 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813278 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 215617 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 566978 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684014 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 425595 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 714766 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 400269 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204814 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 745261 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 886319 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916417 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 756985 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 850149 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854208 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 938551 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553915 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 232048 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 652843 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 507290 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527162 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230432 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 441034 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535514 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 843078 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 508881 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 257492 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496455 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 632284 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 214351 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 512031 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560149 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 693490 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 877780 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 537030 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250352 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 525905 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 148484 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 680617 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681607 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 41 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 451854 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56112 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 33786 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 861295 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 169957 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 927881 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79565 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 888660 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 624004 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 346568 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 485761 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 526496 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 606791 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 759471 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 103123 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 657562 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 453523 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594343 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 431982 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 862130 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 891437 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 555987 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 254541 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 818299 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 820329 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571237 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 366139 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 241538 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71846 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 149004 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 38421 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 141092 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 158553 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 15822 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 77252 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 139199 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 166133 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 50497 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 99951 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 149749 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 129438 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 28854 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 76151 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P154855 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P369825 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P154855 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P369825 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P369825 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P154855 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P369825 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P194097 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 93642 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 62588 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 1638 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 45356 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 39132 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 13406 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 12758 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 96332 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 103639 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 14511 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 57001 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 40383 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 11781 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 86763 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 90161 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 100570 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10907797 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10907800 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10907799 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10907796 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10907801 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10907795 | - | - |
| Rat Genome 230 2.0 Array | 1368974_at | - | - |
| Rat Genome 230 2.0 Array | 1368974_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1368974_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1368974_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0800000009.rn.1 | TC0800000002.rn.1 | RTA TC |
| Rat Expression Set 230 | 1368974_at | - | - |
| Rat Expression Set 230 | 1368974_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10907795 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21598498 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1376307 | - | - |

Gucy1a2
Genomic Map