Assay Details
| Gene Symbol: Birc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023987.3 | NP_076477.3 | 6 - 7 | 1317 | 78 | |
| XM_006242499.2 | XP_006242561.1 | 5 - 6 | 3280 | 78 | ||
| XM_006242500.2 | XP_006242562.1 | 6 - 7 | 2050 | 78 | ||
| XM_006242502.2 | XP_006242564.1 | 5 - 6 | 1549 | 78 | ||
| XM_006242503.2 | XP_006242565.1 | 5 - 6 | 1508 | 78 | ||
| GenBank mRNA | AF183430.1 | - | 4 - 5 | 1085 | 78 | |
| BC083555.1 | - | 6 - 7 | 1317 | 7112819 | 78 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Birc3
Genomic Map