Assay Details
| Gene Symbol: Mdk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006234592.2 | XP_006234654.1 | 1 - 2 | 108 | 73 | |
| GenBank mRNA | AF315950.1 | - | 1 - 2 | 74 | 73 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF315950.1, XM_006234592.2 transcripts.
Set Membership: |
|

Mdk
Genomic Map