Assay Details
Gene Symbol: Ppp1ca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031527.1 | NP_113715.1 | 2 - 3 | 251 | 81 | |
GenBank mRNA | BC070517.1 | - | 2 - 3 | 231 | 7104674 | 81 |
D00859.1 | - | 2 - 3 | 251 | 81 | ||
D90163.1 | - | 2 - 3 | 235 | 81 | ||
FQ224860.1 | - | 2 - 3 | 241 | 81 | ||
FQ231613.1 | - | 2 - 3 | 260 | 81 | ||
FQ232496.1 | - | 2 - 3 | 259 | 81 | ||
FQ233040.1 | - | 2 - 3 | 251 | 81 | ||
FQ235125.1 | - | 2 - 3 | 251 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
