Assay Details
| Gene Symbol: Ppp1ca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031527.1 | NP_113715.1 | 5 - 6 | 812 | 92 | |
| GenBank mRNA | BC070517.1 | - | 5 - 6 | 792 | 7104674 | 92 |
| D00859.1 | - | 5 - 6 | 812 | 92 | ||
| D90163.1 | - | 5 - 6 | 796 | 92 | ||
| FQ224860.1 | - | 5 - 6 | 802 | 92 | ||
| FQ231613.1 | - | 5 - 6 | 821 | 92 | ||
| FQ232496.1 | - | 5 - 6 | 820 | 92 | ||
| FQ233040.1 | - | 5 - 6 | 812 | 92 | ||
| M58437.1 | - | 1 - 2 | 117 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppp1ca
Genomic Map