Assay Details
Gene Symbol: Sult1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031834.1 | NP_114022.1 | 5 - 6 | 568 | 76 | |
GenBank mRNA | AF394783.1 | - | 4 - 5 | 491 | 76 | |
FQ209630.1 | - | 5 - 6 | 577 | 76 | ||
FQ210886.1 | - | 5 - 6 | 573 | 76 | ||
FQ218459.1 | - | 5 - 6 | 573 | 76 | ||
FQ218506.1 | - | 5 - 6 | 582 | 76 | ||
FQ218658.1 | - | 5 - 6 | 572 | 76 | ||
FQ219178.1 | - | 5 - 6 | 573 | 76 | ||
FQ219521.1 | - | 5 - 6 | 573 | 76 | ||
FQ219542.1 | - | 5 - 6 | 573 | 76 | ||
L19998.1 | - | 5 - 6 | 568 | 76 | ||
X52883.1 | - | 5 - 6 | 556 | 76 | ||
X68640.1 | - | 4 - 5 | 473 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
