Assay Details
| Gene Symbol: Dut | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040271.1 | NP_001035361.1 | 2 - 3 | 266 | 77 | |
| NM_053592.2 | NP_446044.1 | 3 - 4 | 386 | 77 | ||
| XM_008762188.1 | XP_008760410.1 | 3 - 4 | 424 | 77 | ||
| GenBank mRNA | CB325379.1 | - | 2 - 3 | 280 | 77 | |
| FQ220451.1 | - | 421 | 77 | |||
| U64030.2 | - | 3 - 4 | 386 | 77 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Dut
Genomic Map