Assay Details
Gene Symbol: Ltc4s | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053639.2 | NP_446091.2 | 2 - 3 | 180 | 62 | |
XM_006246219.2 | XP_006246281.1 | 2 - 3 | 373 | 62 | ||
XM_008767655.1 | XP_008765877.1 | 3 - 4 | 1201 | 62 | ||
GenBank mRNA | AB048790.2 | - | 2 - 3 | 180 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
