Assay Details
Gene Symbol: Mmp16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080776.1 | NP_542954.1 | 4 - 5 | 1138 | 69 | |
XM_006237921.2 | XP_006237983.1 | 4 - 5 | 1204 | 69 | ||
GenBank mRNA | D63886.1 | - | 4 - 5 | 1138 | 69 | |
D85509.1 | - | 4 - 5 | 1138 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
