Assay Details
Gene Symbol: Casp12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130422.1 | NP_569106.1 | 3 - 4 | 414 | 64 | |
XM_008765891.1 | XP_008764113.1 | 4 - 5 | 495 | 64 | ||
GenBank mRNA | AF317633.1 | - | 3 - 4 | 414 | 64 | |
FJ465154.1 | - | 4 - 5 | 477 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
