Assay Details
| Gene Symbol: Fgf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130816.1 | NP_570829.1 | 2 - 3 | 307 | 79 | |
| XM_008767766.1 | XP_008765988.1 | 2 - 3 | 420 | 79 | ||
| GenBank mRNA | AB079263.1 | - | 2 - 3 | 307 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fgf11
Genomic Map