Assay Details
Gene Symbol: Anp32b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131911.2 | NP_571986.1 | 3 - 4 | 497 | 90 | |
XM_008763695.1 | XP_008761917.1 | 4 - 5 | 809 | 90 | ||
GenBank mRNA | AB025581.1 | - | 3 - 4 | 334 | 90 | |
BC086508.1 | - | 3 - 4 | 497 | 7103152 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
