Assay Details
Gene Symbol: Klrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012745.2 | NP_036877.1 | 3 - 4 | 506 | 61 | |
XM_006237078.2 | XP_006237140.1 | 3 - 4 | 624 | 61 | ||
GenBank mRNA | AF009133.1 | - | 3 - 4 | 263 | 61 | |
BC086393.1 | - | 3 - 4 | 506 | 7303354 | 61 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF009133.1, BC086393.1, NM_012745.2, XM_006237078.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
