Assay Details
| Gene Symbol: Spp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012881.2 | NP_037013.2 | 5 - 6 | 327 | 73 | |
| XM_008769996.1 | XP_008768218.1 | 5 - 6 | 400 | 73 | ||
| GenBank mRNA | AB001382.1 | - | 4 - 5 | 221 | 73 | |
| BC078874.1 | - | 5 - 6 | 307 | 7110677 | 73 | |
| CV102664.1 | - | 5 - 6 | 327 | 73 | ||
| FQ213473.1 | - | 5 - 6 | 316 | 73 | ||
| FQ222192.1 | - | 5 - 6 | 343 | 73 | ||
| M14656.1 | - | 5 - 6 | 300 | 73 | ||
| M99252.1 | - | 5 - 6 | 336 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Spp1
Genomic Map