Assay Details
| Gene Symbol: Apobec1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012907.2 | NP_037039.1 | 3 - 4 | 937 | 101 | |
| XM_006237290.2 | XP_006237352.1 | 4 - 5 | 1129 | 101 | ||
| XM_006237291.2 | XP_006237353.1 | 4 - 5 | 1185 | 101 | ||
| XM_006237292.2 | XP_006237354.1 | 4 - 5 | 706 | 101 | ||
| XM_006237293.2 | XP_006237355.1 | 4 - 5 | 861 | 101 | ||
| XM_008763219.1 | XP_008761441.1 | 4 - 5 | 776 | 101 | ||
| XM_008763220.1 | XP_008761442.1 | 4 - 5 | 645 | 101 | ||
| GenBank mRNA | BC085335.1 | - | 3 - 4 | 937 | 7301703 | 101 |
| L07114.1 | - | 3 - 4 | 473 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Apobec1
Genomic Map