Assay Details
| Gene Symbol: Ggh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012960.2 | NP_037092.1 | 5 - 6 | 652 | 71 | |
| GenBank mRNA | BC087602.1 | - | 5 - 6 | 652 | 7312627 | 71 |
| U38379.1 | - | 5 - 6 | 502 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ggh
Genomic Map