Assay Details
| Gene Symbol: Shox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013028.1 | NP_037160.1 | 3 - 4 | 350 | 107 | |
| GenBank mRNA | AJ002258.1 | - | 3 - 4 | 371 | 107 | |
| AJ002259.1 | - | 3 - 4 | 350 | 107 | ||
| AJ002260.1 | - | 2 - 3 | 323 | 107 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Shox2
Genomic Map