Assay Details
| Gene Symbol: Ugt8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019276.3 | NP_062149.1 | 2 - 3 | 888 | 114 | |
| GenBank mRNA | L21698.1 | - | 2 - 3 | 935 | 114 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ugt8
Genomic Map