Assay Details
Gene Symbol: Ppp6c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133589.2 | NP_598273.2 | 4 - 5 | 478 | 80 | |
GenBank mRNA | BC078747.1 | - | 4 - 5 | 478 | 7131278 | 80 |
X77236.1 | - | 4 - 5 | 443 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
