Assay Details
| Gene Symbol: Adh7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134329.1 | NP_599156.1 | 1 - 2 | 69 | 58 | |
| XM_008761493.1 | XP_008759715.1 | 2 - 3 | 456 | 58 | ||
| GenBank mRNA | X98746.1 | - | 1 - 2 | 69 | 58 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_134329.1, X98746.1 transcripts.
Set Membership: |
|

Adh7
Genomic Map