Assay Details
| Gene Symbol: Adh7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134329.1 | NP_599156.1 | 7 - 8 | 1013 | 75 | |
| XM_008761493.1 | XP_008759715.1 | 8 - 9 | 1400 | 75 | ||
| GenBank mRNA | X98746.1 | - | 7 - 8 | 1013 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Adh7
Genomic Map