Assay Details
| Gene Symbol: Scp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138508.4 | NP_612517.2 | 11 - 12 | 1095 | 70 | |
| GenBank mRNA | BC081713.1 | - | 11 - 12 | 1108 | 7109192 | 70 |
| M34728.1 | - | 5 - 6 | 561 | 70 | ||
| M57453.1 | - | 11 - 12 | 1095 | 70 | ||
| M58287.1 | - | 10 - 11 | 981 | 70 | ||
| M62763.1 | - | 11 - 12 | 1122 | 70 | ||
| X60654.1 | - | 11 - 12 | 1075 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Scp2
Genomic Map