Assay Details
Gene Symbol: Slc16a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138831.1 | NP_620186.1 | 1 - 2 | 527 | 74 | |
XM_008772974.1 | XP_008771196.1 | 1 - 2 | 566 | 74 | ||
XM_008772975.1 | XP_008771197.1 | 1 - 2 | 565 | 74 | ||
GenBank mRNA | AB047324.1 | - | 1 - 2 | 527 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
