Assay Details
Gene Symbol: Cdipt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138899.2 | NP_620254.1 | 3 - 4 | 725 | 73 | |
XM_006230208.1 | XP_006230270.1 | 3 - 4 | 899 | 73 | ||
GenBank mRNA | BC070876.1 | - | 3 - 4 | 725 | 7096385 | 73 |
D82928.1 | - | 3 - 4 | 473 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
