Assay Details
| Gene Symbol: Obp1f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138903.1 | NP_620258.1 | 242 | 103 | ||
| GenBank mRNA | AJ251322.1 | - | 226 | 103 | ||
| J03093.1 | - | 242 | 103 | |||
| Gene Symbol: LOC100362409 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006257194.2 | XP_006257256.1 | 2 - 3 | 493 | 103 | |
| Gene Symbol: LOC103690319 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008773238.1 | XP_008771460.1 | 2 - 3 | 493 | 103 | |
More Information
Set Membership: |
|

Obp1f
Genomic Map