Assay Details
Gene Symbol: Lgals3bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139096.1 | NP_620796.1 | 2 - 3 | 204 | 59 | |
GenBank mRNA | AF065438.1 | - | 2 - 3 | 204 | 59 | |
AY552591.1 | - | 1 - 2 | 66 | 59 | ||
BC081724.1 | - | 2 - 3 | 177 | 7123606 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
