Assay Details
| Gene Symbol: Slc15a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139341.1 | NP_647557.1 | 7 - 8 | 75 | ||
| GenBank mRNA | AB026665.1 | - | 7 - 8 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc15a3
Genomic Map