Assay Details
| Gene Symbol: Fmo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144737.2 | NP_653338.2 | 1 - 2 | 92 | 68 | |
| XM_008769605.1 | XP_008767827.1 | 1 - 2 | 166 | 68 | ||
| GenBank mRNA | AF458414.1 | - | 1 - 2 | 59 | 68 | |
| BC070904.1 | - | 1 - 2 | 92 | 7100688 | 68 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF458414.1, BC070904.1, NM_144737.2, XM_008769605.1 transcripts.
Set Membership: |
|

Fmo2
Genomic Map