Assay Details
| Gene Symbol: Tac4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172328.2 | NP_758831.1 | 2 - 3 | 211 | 68 | |
| GenBank mRNA | AF521561.1 | - | 2 - 3 | 211 | 68 | |
| AY471575.1 | - | 2 - 3 | 211 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tac4
Genomic Map