Assay Details
| Gene Symbol: Prss3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173127.1 | NP_775150.1 | 1 - 2 | 46 | 79 | |
| GenBank mRNA | FQ226997.1 | - | 1 - 2 | 77 | 79 | |
| FQ230379.1 | - | 1 - 2 | 77 | 79 | ||
| FQ235277.1 | - | 1 - 2 | 77 | 79 | ||
| M16624.1 | - | 1 - 2 | 46 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prss3
Genomic Map