Assay Details
| Gene Symbol: Obp2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173148.1 | NP_775171.1 | 336 | 124 | ||
| XM_008761579.1 | XP_008759801.1 | 4 - 5 | 441 | 124 | ||
| GenBank mRNA | M76734.1 | - | 336 | 124 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Obp2b
Genomic Map