Assay Details
| Gene Symbol: Cox6a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012812.3 | NP_036944.2 | 1 - 2 | 144 | 76 | |
| XM_006230218.1 | XP_006230280.1 | 2 - 3 | 193 | 76 | ||
| GenBank mRNA | BM392424.1 | - | 2 - 3 | 76 | ||
| BQ782365.1 | - | 2 - 3 | 76 | |||
| FM058713.1 | - | 2 - 3 | 198 | 76 | ||
| FQ217951.1 | - | 1 - 2 | 94 | 76 | ||
| FQ223608.1 | - | 1 - 2 | 100 | 76 | ||
| FQ224015.1 | - | 1 - 2 | 102 | 76 | ||
| FQ224196.1 | - | 1 - 2 | 101 | 76 | ||
| FQ224467.1 | - | 1 - 2 | 90 | 76 | ||
| FQ224572.1 | - | 1 - 2 | 109 | 76 | ||
| X12554.1 | - | 1 - 2 | 65 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cox6a2
Genomic Map