Assay Details
| Gene Symbol: Npap60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012991.1 | NP_037123.1 | 1 - 2 | 307 | 65 | |
| GenBank mRNA | U41845.2 | - | 1 - 2 | 307 | 65 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_012991.1, U41845.2 transcripts.
Set Membership: |
|

Npap60
Genomic Map