Assay Details
Gene Symbol: Pou1f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013008.3 | NP_037140.2 | 1 - 2 | 259 | 63 | |
GenBank mRNA | BC061563.1 | - | 1 - 2 | 263 | 6918965 | 63 |
L01506.1 | - | 1 - 2 | 227 | 63 | ||
M23253.1 | - | 1 - 2 | 173 | 63 | ||
X12658.1 | - | 1 - 2 | 196 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
