Assay Details
Gene Symbol: Gad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017007.1 | NP_058703.1 | 4 - 5 | 480 | 63 | |
XM_008761878.1 | XP_008760100.1 | 3 - 4 | 288 | 63 | ||
GenBank mRNA | EU791557.1 | - | 2 - 3 | 383 | 63 | |
GQ468528.1 | - | 3 - 4 | 659 | 63 | ||
M34445.1 | - | 4 - 5 | 480 | 63 | ||
M38350.1 | - | 4 - 5 | 397 | 63 | ||
M76177.1 | - | 4 - 5 | 481 | 63 | ||
X57572.1 | - | 4 - 5 | 63 | |||
X57573.1 | - | 4 - 5 | 480 | 63 |

More Information
Additional Information:
This assay is in the 5'UTR region of the EU791557.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
