Assay Details
| Gene Symbol: Glul | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017073.3 | NP_058769.4 | 2 - 3 | 287 | 83 | |
| GenBank mRNA | BC072694.1 | - | 2 - 3 | 287 | 7103110 | 83 |
| FQ213256.1 | - | 2 - 3 | 298 | 83 | ||
| FQ216093.1 | - | 2 - 3 | 292 | 83 | ||
| FQ226911.1 | - | 2 - 3 | 310 | 83 | ||
| FQ227474.1 | - | 2 - 3 | 299 | 83 | ||
| FQ232853.1 | - | 2 - 3 | 348 | 83 | ||
| FQ232963.1 | - | 2 - 3 | 299 | 83 | ||
| FQ233770.1 | - | 2 - 3 | 299 | 83 | ||
| FQ234289.1 | - | 2 - 3 | 300 | 83 | ||
| M29579.1 | - | 2 - 3 | 298 | 83 | ||
| M91652.1 | - | 2 - 3 | 296 | 83 | ||
| X07921.1 | - | 2 - 3 | 298 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Glul
Genomic Map